Product Details

SNP ID
rs4735736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:76691404 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAACATGGTGTGAACAAGAACAAC[A/G]CTTCAATTCCGTGTAAGGAAAGGAT
Phenotype
MIM: 606940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFHX4 PubMed Links

Gene Details

Gene
ZFHX4
Gene Name
zinc finger homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024721.4 Intron NP_078997.4
XM_011517592.2 Intron XP_011515894.1
XM_011517593.2 Intron XP_011515895.1
XM_011517594.2 Intron XP_011515896.1
XM_011517595.2 Intron XP_011515897.1
XM_011517596.2 Intron XP_011515898.1
XM_011517597.2 Intron XP_011515899.1
XM_017013845.1 Intron XP_016869334.1
Gene
ZFHX4-AS1
Gene Name
ZFHX4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details