Product Details

SNP ID
rs12938530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75117396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCAAAAGTGAGGCCTTTATTG[G/T]GGGGCGACAAGCAAGAAGAATCCGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARMC7 PubMed Links

Gene Details

Gene
ARMC7
Gene Name
armadillo repeat containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304271.1 Intron NP_001291200.1
NM_001304272.1 Intron NP_001291201.1
NM_001304273.1 Intron NP_001291202.1
NM_024585.3 Intron NP_078861.1

View Full Product Details