Product Details

SNP ID
rs1937236
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:192165317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTCCTGTTAAGATGTTTATCGA[C/T]GACAATGCTTGCACAGTGGGACTTG
Phenotype
MIM: 607192
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS18 PubMed Links

Gene Details

Gene
RGS18
Gene Name
regulator of G-protein signaling 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130782.2 Intron NP_570138.1
XM_011509891.2 Intron XP_011508193.1

View Full Product Details