Product Details

SNP ID
rs4357510
Assay Type
Functionally tested
NCBI dbSNP Submissions
57
Location
Chr.1:192643264 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTTGGCACATAACAAGTATTCAA[A/C]AGACTTTTGAATGAATCGTAAAGAC
Phenotype
MIM: 607190
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RGS13 PubMed Links

Gene Details

Gene
RGS13
Gene Name
regulator of G-protein signaling 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002927.4 Intron NP_002918.1
NM_144766.2 Intron NP_658912.1

View Full Product Details