Product Details

SNP ID
rs13136658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:37462649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACCACACTTCAAATTTCAAATG[A/G]TGTGAGCAATTTTGCCGTCCATTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C4orf19 PubMed Links

Gene Details

Gene
C4orf19
Gene Name
chromosome 4 open reading frame 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104629.1 Intron NP_001098099.1
NM_018302.2 Intron NP_060772.2
XM_011513712.2 Intron XP_011512014.1
XM_011513713.2 Intron XP_011512015.1
XM_017008361.1 Intron XP_016863850.1

View Full Product Details