Product Details
- SNP ID
-
rs12737747
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:20189315 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTCACAGTGGTGGTCAGGATTCA[A/G]AGGGAGGCAGGGAGCCCAATAAGAT
- Phenotype
-
MIM: 616783
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
UBXN10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117788762] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UBXN10
- Gene Name
- UBX domain protein 10
- Gene
- UBXN10-AS1
- Gene Name
- UBXN10 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details