Product Details

SNP ID
rs7133651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21017123 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCAGTATTACACCTTTTTACTT[A/G]AGGAACTTGAACATCCACAAATCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLCO1B7 PubMed Links

Gene Details

Gene
SLCO1B7
Gene Name
solute carrier organic anion transporter family member 1B7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009562.4 Intron NP_001009562.3

View Full Product Details