Product Details

SNP ID
rs12815446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68298692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATAAGACCTCACTGCATTTAA[C/G]TGAGAGCTCCCCCAGCCACCTTCAT
Phenotype
MIM: 613813
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MDM1 PubMed Links

Gene Details

Gene
MDM1
Gene Name
Mdm1 nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205028.1 Intron NP_001191957.1
NM_001205029.1 Intron NP_001191958.1
NM_017440.4 Intron NP_059136.2
NM_020128.2 Intron NP_064513.1
XM_005269026.1 Intron XP_005269083.1
XM_011538569.1 Intron XP_011536871.1
XM_017019646.1 Intron XP_016875135.1
XM_017019647.1 Intron XP_016875136.1
XM_017019648.1 Intron XP_016875137.1
XM_017019649.1 Intron XP_016875138.1
XM_017019650.1 Intron XP_016875139.1

View Full Product Details