Product Details
- SNP ID
-
rs12302426
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:72096124 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGATTCCTTAAGATAAATCTTTT[C/G]TTATGTGTATACACACACACACACA
- Phenotype
-
MIM: 606950
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TRHDE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144560604] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TRHDE
- Gene Name
- thyrotropin releasing hormone degrading enzyme
View Full Product Details