Product Details

SNP ID
rs11058594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122527884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCCTTCCCTTCCCAAACAAAT[C/T]CTTCTTCAGCGACGCCTCACCTTAT
Phenotype
MIM: 607363
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KNTC1 PubMed Links

Gene Details

Gene
KNTC1
Gene Name
kinetochore associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014708.4 Intron NP_055523.1
XM_006719706.2 Intron XP_006719769.1
XM_011539027.2 Intron XP_011537329.1
XM_011539029.2 Intron XP_011537331.1
XM_011539030.1 Intron XP_011537332.1
XM_017020254.1 Intron XP_016875743.1
XM_017020255.1 Intron XP_016875744.1
XM_017020256.1 Intron XP_016875745.1
XM_017020257.1 Intron XP_016875746.1
Gene
RSRC2
Gene Name
arginine and serine rich coiled-coil 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023012.5 Intron NP_075388.2
XM_005253601.1 Intron XP_005253658.1
XM_005253602.2 Intron XP_005253659.1
XM_005253604.1 Intron XP_005253661.1
XM_011538688.1 Intron XP_011536990.1
XM_011538689.1 Intron XP_011536991.1
XM_017019830.1 Intron XP_016875319.1
XM_017019831.1 Intron XP_016875320.1
XM_017019832.1 Intron XP_016875321.1
XM_017019833.1 Intron XP_016875322.1

View Full Product Details