Product Details

SNP ID
rs6488898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123719285 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGCCACTTCTTTTATCTCTAAA[A/G]TCTTTACCATCTTAGTCTGGTACTA
Phenotype
MIM: 611716
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0A2 PubMed Links

Gene Details

Gene
ATP6V0A2
Gene Name
ATPase H+ transporting V0 subunit a2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012463.3 Intron NP_036595.2

View Full Product Details