Product Details

SNP ID
rs11649604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19531183 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTTATCAGCTATTTGATAACAC[A/G]TTGCTTGATTTAATGTGGACCAAAA
Phenotype
MIM: 609544 MIM: 605943
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCP110 PubMed Links

Gene Details

Gene
CCP110
Gene Name
centriolar coiled-coil protein 110
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199022.2 Intron NP_001185951.2
NM_001323569.1 Intron NP_001310498.1
NM_001323570.1 Intron NP_001310499.1
NM_001323571.1 Intron NP_001310500.1
NM_001323572.1 Intron NP_001310501.1
NM_001323576.1 Intron NP_001310505.1
NM_001323577.1 Intron NP_001310506.1
NM_014711.5 Intron NP_055526.4
XM_011545991.2 Intron XP_011544293.1
XM_011545992.2 Intron XP_011544294.1
XM_017023908.1 Intron XP_016879397.1
Gene
GDE1
Gene Name
glycerophosphodiester phosphodiesterase 1
There are no transcripts associated with this gene.

View Full Product Details