Product Details

SNP ID
rs13428042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27290257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAATGAAGAAAGCTTTGGAAACA[A/C]TGGAATATAATCACCAAAATATTTA
Phenotype
MIM: 613946 MIM: 606474
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC5G PubMed Links

Gene Details

Gene
DNAJC5G
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
There are no transcripts associated with this gene.

Gene
TRIM54
Gene Name
tripartite motif containing 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032546.3 Intron NP_115935.3
NM_187841.2 Intron NP_912730.2

View Full Product Details