Product Details

SNP ID
rs9333640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:155802522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTTGGCCTCACAAAATAATCTTT[T/C]CCCCGTGAGTACAGTTCACATGATC
Phenotype
MIM: 600725
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SHH PubMed Links

Gene Details

Gene
SHH
Gene Name
sonic hedgehog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000193.3 1918 UTR 3 NP_000184.1
NM_001310462.1 1918 Intron NP_001297391.1
XM_011516479.2 1918 Intron XP_011514781.1
XM_011516480.2 1918 Intron XP_011514782.1

View Full Product Details