Product Details

SNP ID
rs9333634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:155803659 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGGTCCTTCACCAGCTTGGTGCC[A/G]CCCTGCTCCAGGTGCACCGTGGCCG
Phenotype
MIM: 600725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SHH PubMed Links

Gene Details

Gene
SHH
Gene Name
sonic hedgehog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000193.3 3579 Silent Mutation GGC,GGT G210G NP_000184.1
NM_001310462.1 3579 Intron NP_001297391.1
XM_011516479.2 3579 Silent Mutation GGC,GGT G123G XP_011514781.1
XM_011516480.2 3579 Silent Mutation GGC,GGT G123G XP_011514782.1

View Full Product Details