Product Details

SNP ID
rs13127570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169625304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTTTCTTTTTAGCTTAACAGA[A/G]AGTTGACTGACACACCTTTGGGAAT
Phenotype
MIM: 600580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCN3 PubMed Links

Gene Details

Gene
CLCN3
Gene Name
chloride voltage-gated channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243372.1 Intron NP_001230301.1
NM_001243374.1 Intron NP_001230303.1
NM_001829.3 Intron NP_001820.2
NM_173872.3 Intron NP_776297.2
XM_005262726.2 Intron XP_005262783.1
XM_011531586.2 Intron XP_011529888.1

View Full Product Details