Product Details

SNP ID
rs13435339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70365524 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTATGAACATCATTTTCCTGAT[A/G]AGAAAAAGAGACTCAAAAAGGATTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMR3A PubMed Links

Gene Details

Gene
SMR3A
Gene Name
submaxillary gland androgen regulated protein 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012390.3 Intron NP_036522.3
XM_017008021.1 Intron XP_016863510.1

View Full Product Details