Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130016.2 | 922 | Intron | NP_001123488.1 | ||
NM_001130017.2 | 922 | Intron | NP_001123489.1 | ||
NM_001179.5 | 922 | Intron | NP_001170.2 | ||
XM_005262997.1 | 922 | Intron | XP_005263054.1 | ||
XM_005262999.1 | 922 | Intron | XP_005263056.1 | ||
XM_005263003.1 | 922 | Intron | XP_005263060.1 | ||
XM_005263004.1 | 922 | Intron | XP_005263061.1 | ||
XM_006714220.1 | 922 | Intron | XP_006714283.1 | ||
XM_011531971.1 | 922 | Intron | XP_011530273.1 | ||
XM_017008206.1 | 922 | Intron | XP_016863695.1 | ||
XM_017008207.1 | 922 | Intron | XP_016863696.1 | ||
XM_017008208.1 | 922 | Intron | XP_016863697.1 | ||
XM_017008209.1 | 922 | Intron | XP_016863698.1 | ||
XM_017008210.1 | 922 | Intron | XP_016863699.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001302123.1 | 922 | UTR 3 | NP_001289052.1 | ||
NM_005409.4 | 922 | UTR 3 | NP_005400.1 |