Product Details

SNP ID
rs12737795
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:77096499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTTGGCTTGCTTCTGCTCTCTCT[A/C]AGACCTCTTCCACCACCAGGAGAAA
Phenotype
MIM: 605087
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PIGK PubMed Links

Gene Details

Gene
PIGK
Gene Name
phosphatidylinositol glycan anchor biosynthesis class K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005482.2 Intron NP_005473.1

View Full Product Details