Product Details
- SNP ID
-
rs12757862
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:68474802 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTCCTTCCTCATCAAATTTTCCT[G/T]CTTGACTAGATTAAAAAATTTCAAC
- Phenotype
-
MIM: 612002
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DEPDC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75750431] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DEPDC1
- Gene Name
- DEP domain containing 1
View Full Product Details