Product Details

SNP ID
rs12039939
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:32664150 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACCGTGTTAGCCAGCATAGTCTT[G/T]ATCTCCTGAACCCTGTCAAATTTAA
Phenotype
MIM: 602923
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RBBP4 PubMed Links

Gene Details

Gene
RBBP4
Gene Name
RB binding protein 4, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135255.1 Intron NP_001128727.1
NM_001135256.1 Intron NP_001128728.1
NM_005610.2 Intron NP_005601.1

View Full Product Details