Product Details

SNP ID
rs7279340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45412086 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGGGAATGCCGGGTTTGTGGCT[C/T]GTGCCCACTGCCCTGCAGGTGCATT
Phenotype
MIM: 120328
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL18A1 PubMed Links

Gene Details

Gene
COL18A1
Gene Name
collagen type XVIII alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030582.3 Intron NP_085059.2
NM_130444.2 Intron NP_569711.2
NM_130445.3 Intron NP_569712.2
Gene
COL18A1-AS1
Gene Name
COL18A1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
COL18A1-AS2
Gene Name
COL18A1 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details