Product Details

SNP ID
rs12621841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:163606268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATTTGGGAGAGAAGCATGCCA[C/G]AAGTGTAGAGTATTTCCATTTGTGT
Phenotype
MIM: 605295
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FIGN PubMed Links

Gene Details

Gene
FIGN
Gene Name
fidgetin, microtubule severing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321825.1 5879 UTR 3 NP_001308754.1
NM_018086.3 5879 UTR 3 NP_060556.2
XM_011511389.2 5879 UTR 3 XP_011509691.1
XM_017004419.1 5879 UTR 3 XP_016859908.1

View Full Product Details