Product Details

SNP ID
rs13215652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158247605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTTGAGTCAGTTATACCTCTG[A/T]TTTGTGTGTGTGAATGTTTTTCAGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TULP4 PubMed Links

Gene Details

Gene
TULP4
Gene Name
tubby like protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007466.2 Intron NP_001007467.1
NM_020245.4 Intron NP_064630.2
XM_011535946.1 Intron XP_011534248.1
XM_017011063.1 Intron XP_016866552.1
XM_017011064.1 Intron XP_016866553.1
XM_017011065.1 Intron XP_016866554.1
XM_017011066.1 Intron XP_016866555.1
XM_017011067.1 Intron XP_016866556.1
XM_017011068.1 Intron XP_016866557.1
XM_017011069.1 Intron XP_016866558.1
XM_017011070.1 Intron XP_016866559.1
XM_017011071.1 Intron XP_016866560.1

View Full Product Details