Product Details

SNP ID
rs11953417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115804975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAAAATAAAATCTACAGATTC[A/T]CAAAATTATCTTCCAATATTAAGAA
Phenotype
MIM: 603943
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDO1 PubMed Links

Gene Details

Gene
CDO1
Gene Name
cysteine dioxygenase type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323565.1 1372 UTR 3 NP_001310494.1
NM_001323566.1 1372 UTR 3 NP_001310495.1
NM_001323567.1 1372 UTR 3 NP_001310496.1
NM_001801.2 1372 UTR 3 NP_001792.2

View Full Product Details