Product Details

SNP ID
rs11466298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70448266 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAGTCACAGCCTGTAGAGCA[C/G]TGGGAAAAATCCAGGCTCCATAAGA
Phenotype
MIM: 190170
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TGFA PubMed Links

Gene Details

Gene
TGFA
Gene Name
transforming growth factor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099691.2 3488 UTR 3 NP_001093161.1
NM_001308158.1 3488 UTR 3 NP_001295087.1
NM_001308159.1 3488 UTR 3 NP_001295088.1
NM_003236.3 3488 UTR 3 NP_003227.1

View Full Product Details