Product Details

SNP ID
rs12052667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20555343 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCCGGCTGCCTAGTTTTTGGGGC[C/G]CAGGGCAGAAGGAAACTGTGGGGTC
Phenotype
MIM: 609359
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HS1BP3 PubMed Links

Gene Details

Gene
HS1BP3
Gene Name
HCLS1 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022460.3 Intron NP_071905.3
XM_017004696.1 Intron XP_016860185.1
XM_017004697.1 Intron XP_016860186.1
XM_017004698.1 Intron XP_016860187.1
XM_017004699.1 Intron XP_016860188.1
XM_017004700.1 Intron XP_016860189.1
XM_017004701.1 Intron XP_016860190.1

View Full Product Details