Product Details

SNP ID
rs9658627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53103938 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCCCTGAGTGCCCTGTTCCCAG[C/T]AATGTCACCAGGCCCCCTGCTTGCC
Phenotype
MIM: 146735 MIM: 601311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFBP6 PubMed Links

Gene Details

Gene
IGFBP6
Gene Name
insulin like growth factor binding protein 6
There are no transcripts associated with this gene.

Gene
SOAT2
Gene Name
sterol O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003578.3 Intron NP_003569.1
XM_011538849.2 Intron XP_011537151.1
XM_011538850.2 Intron XP_011537152.1
XM_011538851.2 Intron XP_011537153.1

View Full Product Details