Product Details

SNP ID
rs3816152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53976267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTTCTTTCCACCTGGAGGGAAAG[C/G]GGGGGACGCCCCCAGTGAGATTCAA
Phenotype
MIM: 611400 MIM: 605560 MIM: 605559
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HOTAIR PubMed Links

Gene Details

Gene
HOTAIR
Gene Name
HOX transcript antisense RNA
There are no transcripts associated with this gene.

Gene
HOXC-AS3
Gene Name
HOXC cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXC10
Gene Name
homeobox C10
There are no transcripts associated with this gene.

Gene
HOXC11
Gene Name
homeobox C11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014212.3 1903 UTR 3 NP_055027.1

View Full Product Details