Product Details
- SNP ID
-
rs12830806
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:69360808 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCACCGCGCCCAGCCGTGGTTCAT[A/T]CTTTTTAAAGAAAAATTTCCTTTCC
- Phenotype
-
MIM: 153450
MIM: 602116
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LYZ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs552180897] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LYZ
- Gene Name
- lysozyme
There are no transcripts associated with this gene.
- Gene
- YEATS4
- Gene Name
- YEATS domain containing 4
View Full Product Details