Product Details
- SNP ID
-
rs12810681
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:69248304 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAAAAATACATTTAGTCCTGCTG[G/T]TAATAACCCTTCCATTCTTTTTTAC
- Phenotype
-
MIM: 604979
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CPSF6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75737671] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPSF6
- Gene Name
- cleavage and polyadenylation specific factor 6
View Full Product Details