Product Details

SNP ID
rs11614988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69751753 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATTAGGGATATATATTTATGTT[G/T]TTATGTTTTAAAATCACTCGAAGTA
Phenotype
MIM: 608686
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB3IP PubMed Links

Gene Details

Gene
RAB3IP
Gene Name
RAB3A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024647.3 Intron NP_001019818.1
NM_001278402.1 Intron NP_001265331.1
NM_022456.4 Intron NP_071901.2
NM_175623.3 Intron NP_783322.1
NM_175624.3 Intron NP_783323.1
NM_175625.3 Intron NP_783324.1
XM_006719226.3 Intron XP_006719289.3
XM_006719227.3 Intron XP_006719290.3
XM_017018773.1 Intron XP_016874262.1
XM_017018774.1 Intron XP_016874263.1
XM_017018775.1 Intron XP_016874264.1
XM_017018776.1 Intron XP_016874265.1
XM_017018777.1 Intron XP_016874266.1

View Full Product Details