Product Details

SNP ID
rs11177889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69832877 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAATCTGTTTAGGATGATGCCTAG[G/T]TGTTGGTGTCAACAATTCATGGATG
Phenotype
MIM: 608686
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYRFL PubMed Links

Gene Details

Gene
MYRFL
Gene Name
myelin regulatory factor-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182530.2 Intron NP_872336.2
XM_011539052.2 Intron XP_011537354.1
XM_011539055.1 Intron XP_011537357.1
XM_017018967.1 Intron XP_016874456.1
XM_017018968.1 Intron XP_016874457.1
XM_017018969.1 Intron XP_016874458.1
XM_017018970.1 Intron XP_016874459.1
XM_017018971.1 Intron XP_016874460.1
XM_017018972.1 Intron XP_016874461.1
XM_017018973.1 Intron XP_016874462.1
XM_017018974.1 Intron XP_016874463.1
XM_017018975.1 Intron XP_016874464.1
XM_017018976.1 Intron XP_016874465.1
Gene
RAB3IP
Gene Name
RAB3A interacting protein
There are no transcripts associated with this gene.

View Full Product Details