Product Details

SNP ID
rs11104698
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:87982825 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAAAAAAAATGCATCTTAAC[A/G]TATTACATGATTGCTGCAATTACTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C12orf50 PubMed Links

Gene Details

Gene
C12orf50
Gene Name
chromosome 12 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152589.1 Intron NP_689802.1
XM_011537985.1 Intron XP_011536287.1
XM_017018886.1 Intron XP_016874375.1
XM_017018887.1 Intron XP_016874376.1
XM_017018888.1 Intron XP_016874377.1

View Full Product Details