Product Details

SNP ID
rs11067227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109553880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGCAGCAGCAAGGGTGACATTGC[C/T]ACTGACGGGGGCTTCCGAACTGGGG
Phenotype
MIM: 607568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMAB PubMed Links

Gene Details

Gene
MMAB
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblB type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052845.3 3974 UTR 3 NP_443077.1
XM_011538267.2 3974 Intron XP_011536569.1
XM_011538268.2 3974 Intron XP_011536570.1
XM_011538269.2 3974 Intron XP_011536571.1

View Full Product Details