Product Details
- SNP ID
-
rs6766319
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:15429273 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTACTTTGGGAGGCCGAGGTCG[A/G]CAGATAGCTTGAGCCCAGGAGTTTG
- Phenotype
-
MIM: 608315
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EAF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116596727] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EAF1
- Gene Name
- ELL associated factor 1
- Gene
- METTL6
- Gene Name
- methyltransferase like 6
There are no transcripts associated with this gene.
View Full Product Details