Product Details

SNP ID
rs11920912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58010179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGTTCTGGGTCTGGATCTCTTC[C/G]TAAGGAGAACCAGGGACTGGCCCTG
Phenotype
MIM: 603381
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLNB PubMed Links

Gene Details

Gene
FLNB
Gene Name
filamin B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164317.1 Intron NP_001157789.1
NM_001164318.1 Intron NP_001157790.1
NM_001164319.1 Intron NP_001157791.1
NM_001457.3 Intron NP_001448.2
XM_005264978.1 Intron XP_005265035.1

View Full Product Details