Product Details
- SNP ID
-
rs10305852
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:147482505 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGATTCAAAGCACAGCCTGACTCA[G/T]ATTCCTGGAAATGAGCTACAGACCC
- Phenotype
-
MIM: 131243
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EDNRA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10305853] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EDNRA
- Gene Name
- endothelin receptor type A
View Full Product Details