Product Details

SNP ID
rs6816298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:64278333 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCCGCAAGAATTTGAACTAATG[A/G]CATAATACCTATGACACATCTCACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TECRL PubMed Links

Gene Details

Gene
TECRL
Gene Name
trans-2,3-enoyl-CoA reductase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010874.4 2944 Intron NP_001010874.2
XM_005265662.4 2944 UTR 3 XP_005265719.1
XM_005265663.2 2944 Intron XP_005265720.1
XM_005265664.2 2944 Intron XP_005265721.1
XM_005265665.3 2944 Intron XP_005265722.1
XM_017007959.1 2944 Intron XP_016863448.1
XM_017007960.1 2944 Intron XP_016863449.1

View Full Product Details