Product Details

SNP ID
rs11728600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:105553519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTAGGTGATTATTTCCTTTGAGT[A/T]AATTGAATAATACTCAAATATGGTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF38 PubMed Links

Gene Details

Gene
ARHGEF38
Gene Name
Rho guanine nucleotide exchange factor 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242729.1 Intron NP_001229658.1
NM_017700.1 Intron NP_060170.1
XM_006714244.3 Intron XP_006714307.1
XM_011532050.2 Intron XP_011530352.1
XM_011532051.2 Intron XP_011530353.1
Gene
ARHGEF38-IT1
Gene Name
ARHGEF38 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details