Product Details
- SNP ID
-
rs11690262
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:174435245 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCACATCCATAGATTCAATCAAC[C/T]GCAGATAAAAAGATGTAGTCAGGCT
- Phenotype
-
MIM: 614967
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPR155
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79631467] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPR155
- Gene Name
- G protein-coupled receptor 155
- Gene
- SCRN3
- Gene Name
- secernin 3
There are no transcripts associated with this gene.
View Full Product Details