Product Details
- SNP ID
-
rs12119116
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
21
- Location
-
Chr.1:78895189 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGATTAAAACATGTTTATCAAAAGG[A/C]GTTATAGTTTCATGTTATCCAAAAA
- Phenotype
-
MIM: 616419
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79589485] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADGRL4
- Gene Name
- adhesion G protein-coupled receptor L4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022159.3 |
|
Intron |
|
|
NP_071442.2 |
View Full Product Details