Product Details

SNP ID
rs12121914
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:78047931 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTTCTGATTCCTAAGCTAGTGAG[C/G]TTGTTAGCATACTTAACTGACAACT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GIPC2 PubMed Links

Gene Details

Gene
GIPC2
Gene Name
GIPC PDZ domain containing family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304725.1 Intron NP_001291654.1
NM_017655.5 Intron NP_060125.4

View Full Product Details