Product Details

SNP ID
rs12037056
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:66757978 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGGCCTGCAAATCCAAAAATAT[C/G]TGCTATTTGGCCTTTTACAGAAATG
Phenotype
MIM: 611540
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SGIP1 PubMed Links

Gene Details

Gene
SGIP1
Gene Name
SH3 domain GRB2 like endophilin interacting protein 1
There are no transcripts associated with this gene.

Gene
TCTEX1D1
Gene Name
Tctex1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152665.2 Intron NP_689878.2
XM_011540933.2 Intron XP_011539235.1
XM_017000566.1 Intron XP_016856055.1

View Full Product Details