Product Details

SNP ID
rs12043763
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:63375709 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTTTGTTCCTTAGATACTTGGTA[A/G]AACTTGATAGTAAAGGTATTTGGGC
Phenotype
MIM: 604566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG6 PubMed Links

Gene Details

Gene
ALG6
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013339.3 Intron NP_037471.2

View Full Product Details