Product Details
- SNP ID
-
rs10889417
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
72
- Location
-
Chr.1:63370230 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGTATTTTTTTCTCACGATTTGGG[A/G]TTATTCTTAATGATTCTTAAAAAGG
- Phenotype
-
MIM: 604566
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALG6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111396360,rs71244580] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALG6
- Gene Name
- ALG6, alpha-1,3-glucosyltransferase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013339.3 |
|
Intron |
|
|
NP_037471.2 |
View Full Product Details