Product Details

SNP ID
rs11247865
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:26191282 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTGAAGGAAGGTCCTGCCCTCT[C/T]CCAGGACGCCTGGGACATGCAGGAG
Phenotype
MIM: 609121 MIM: 603272
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CATSPER4 PubMed Links

Gene Details

Gene
CATSPER4
Gene Name
cation channel sperm associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198137.1 Intron NP_937770.1
XM_011541432.2 Intron XP_011539734.1
XM_011541433.2 Intron XP_011539735.1
Gene
CNKSR1
Gene Name
connector enhancer of kinase suppressor of Ras 1
There are no transcripts associated with this gene.

View Full Product Details