Product Details

SNP ID
rs12096733
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:26056678 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGAGGCCTCCCTGCCATGGATC[C/T]ATATGTAATTTATGAAGCAGGAATT
Phenotype
MIM: 606131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM63 PubMed Links

Gene Details

Gene
TRIM63
Gene Name
tripartite motif containing 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032588.3 Intron NP_115977.2
XM_017002559.1 Intron XP_016858048.1

View Full Product Details