Product Details

SNP ID
rs11247735
Assay Type
Functionally tested
NCBI dbSNP Submissions
57
Location
Chr.1:25804968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTAATGTTATTATCTGCCTCCCT[A/G]GGCTATCCTGAAACTAAAATGCGAT
Phenotype
MIM: 606210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPN1 PubMed Links

Gene Details

Gene
SEPN1
Gene Name
selenoprotein N, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020451.2 Intron NP_065184.2
NM_206926.1 Intron NP_996809.1

View Full Product Details