Product Details

SNP ID
rs12173116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:168661867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTTATTCCATACTGGGGAAGTG[C/G]CACTATAACATTGTTTTAAAAAATC
Phenotype
MIM: 603745
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLIT3 PubMed Links

Gene Details

Gene
SLIT3
Gene Name
slit guidance ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271946.1 9579 UTR 3 NP_001258875.1
NM_003062.3 9579 UTR 3 NP_003053.1
XM_017009779.1 9579 UTR 3 XP_016865268.1

View Full Product Details